Learn More
ATCC LTR228 Cell Line (ATCC HB-8502) - Human B-cell, derivative of WIL-2NS
Supplier:  ATCC LTR228
Encompass
LTR228 is a human B-cell lymphoid cell line reported as a derivative of the WIL-2NS parent line. It is used in immunology research and as a fusion partner for hybridoma production. The cell line is associated with ATCC catalog HB-8502.
  • Human B-cell origin.
  • Derived from WIL-2NS parent line.
  • Reported tissue of origin: spleen.
  • Common application: fusion partner for human B lymphocytes and immunology research.
  • Synonyms include LTR 228 and LTR 228-TG.
Catalog No. NC3803502