Learn More
Beckman Coulter FisherbrandTM Disposable Microscope Slides, Frosted, 25 x 75 x 1mm, Pack of 72
Supplier:  Beckman Coulter 883371
Encompass
The FisherbrandTM NC1808632 is a durable polypropylene rack for 32 microcentrifuge tubes of 1.5mL. It features an alpha-numeric grid for sample identification and withstands -20°C to 121°C. Dimensions are 12.7cm x 8.3cm x 5.4cm. It ensures organized storage and safe centrifugation, maintaining tube position and orientation.
Catalog No. NC1808632