Learn More
Essendant LUBRICANTRED GREASE
Supplier:  Essendant AMR1003057
Encompass_Preferred
Protect your equipment from acid salt spray and water contaminants indoors and out even under extremely high temperatures Thick red grease withstands heavy shock and impact pressures up to 65 Timken OK Loads Once applied multipurpose lubricant displays excellent adhesive and cohesive properties Extreme anti-wear additives ensure reliable hassle-free results Application- Automotive Maintenance Machinery Moving Parts Applicable Material- Metal Steel Volume Capacity- 14 oz Product Packaging- Tube
Catalog No. 50-000-22990