Learn More
Amsbio LLC CMV-Luciferase (firefly)-2A-GFP (Neo)
Supplier:  Amsbio LLC LVP403PBS
Encompass
Ready-to-use lentivirus / lentiviral particles expressing individually Firefly luciferase and GFP through a bicistronic system (Neomycin marker)
Catalog No. 50-204-1408