Learn More
Medchemexpress LLC Lucialdehyde A | 420781-84-6 | 98.9% | 438.69 g/mol | C30H46O2 | 1MG

Supplier: Medchemexpress LLC HY1254431MG
Lucialdehyde A is a lanostane-type triterpene aldehyde isolated from the fruiting bodies of Ganoderma lucidum. It has reported cytotoxic activity against several tumor cell lines and is provided for research use in small quantities.
- Lanostane-type triterpene aldehyde isolated from Ganoderma lucidum.
- Reported cytotoxic activity against Lewis lung carcinoma, T-47D, Sarcoma 180, and Meth-A cell lines.
- High purity: 98.9%.
- Molecular weight: 438.69 g/mol; formula C30H46O2.
- Solubility: DMSO ≈17.38 mg/mL; warming and sonication may be required.
- Storage: store at 4°C protected from light; in solvent, freeze at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.