Learn More
Medchemexpress LLC Lucifer yellow CH (dipotassium salt) | 71206-95-6 | MFCD00013445 | 99.8% | 521.57 | C13H9K2N5O9S2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW12771550MG
Encompass_Preferred
Lucifer Yellow CH (dipotassium salt) is a high-intensity fluorescent probe containing free hydrazyl groups. It is used as a biological tracer for monitoring neuronal branching and regeneration, detecting and characterizing gap junctions, and performing selective cell ablation after aldehyde fixation. The dye has an approximate excitation/emission of 430 nm/540 nm and is supplied as a solid for preparation of stock solutions.
  • High fluorescence intensity for sensitive detection.
  • Suitable for neuronal tracing and regeneration studies.
  • Enables gap junction detection and characterization.
  • Reacts with fatty aldehydes, allowing post-fixation labeling.
  • Supplied as a solid and available as a DMSO solution for convenience.
  • Stock solutions should be stored protected from light and avoid repeated freeze-thaw cycles.
Catalog No. 50-002-82604