Learn More
Medchemexpress LLC Luciferase-IN-1 | 10205-56-8 | 99.6% | 254.35 | 50 MG

Supplier: Medchemexpress LLC HY13670650MG
Luciferase-IN-1 is a luciferase inhibitor.
- Appears as a solid, light yellow to brown.
- Soluble in DMSO at 10 mg/mL (39.32 mM).
- Multiple in vivo dissolution protocols are available.
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years.
- Recommended storage for solution: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.