Learn More
Creative Biomart Active Recombinant Firefly Luciferase Protein (1-550), N-His-tagged, 1mg
Supplier:  Creative Biomart LUCIFERASE787F1MG
Encompass
Luciferase is a generic term for the class of oxidative enzymes used in bioluminescence and is distinct from a photoprotein. Luciferase catalyzes a bioluminescent reaction which requires the substrate luciferin as well as Mg2+ and ATP, produces green light with a wavelength of 562 nm. Luciferase from firefly is widely used as a reporter for studying gene regulation and function, and for pharmaceutical screening.
Catalog No. NC3308050