Learn More
Enzo Life Sciences Lucigenin, (50mg), CAS Number: 2315-97-1
SDP
Supplier:  Enzo Life Sciences ALX620061M050
Encompass_Preferred
ROS dye . Chemiluminescent probe for the detection of superoxide in biological systems. Alternative Name: bis-N-Methylacridinium nitrate, N,N'-Dimethyl-9,9'-biacridinium dinitrate. Formula: C28H22N4O6. MW: 510.5. CAS Number: 2315-97-1. Purity: ≥97% (HPLC). Appearance: Off-white to brown powder. Solubility: Soluble in DMSO (~12mg/ml), 100% ethanol (~0.5mg/ml) or acetic acid (~10mg/ml, heat up solution). Long Term Storage: Ambient
Catalog No. NC9149496