Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Male Luer Lock to Threaded Adapter, 10-32; 25/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4551884
Encompass
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
Catalog No. NC0883054