Learn More
Vici Precision Sampling, Inc. LUER NEEDLE SIZE 2IN /3PK
Supplier:  Vici Precision Sampling, Inc. 943062
Encompass_Preferred
Luer Needles A-2,C,& D.028 in. x .012 in. x 2 in.(22 gauge) side port taper pkg/3
Catalog No. NC9669741