Learn More
Access Technologies Luer Stub 23 Gauge Blunt Needle Adaptors, 50 per Pack
Supplier:  Access Technologies LSA23
Encompass
23 gauge blunt needle adaptors 50/pk. Lt Blue - Outer Diameter Inches/MM 0.024/0.63; Inner Diameter Inches/MM 0.012/0.31.
Catalog No. NC9400183
 Restricted Item