Learn More
AVANTOR MASTERFLEX (VWR LLC) Luer Adapters, Male Luer Lock to 1/16"" ID, Wide-Bore, Nylon, 25/Pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4550531
Encompass
Male Luer Adapters, Lock to 1/16"ID, Wide-Bore, Nylon, 25/Pk, offers a variety of barbed luer adapters to meet your application's needsStandard hose barb for use with flexible tubingSmooth bore hose barb to reduce flow turbulence and risk of line contaminationWide-bore hose barb for enhanced fluid path flowLow-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
Catalog No. NC1108514