Learn More
Medchemexpress LLC LUF5771 | 1141802-49-4 | 99.49% | 357.44 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13930350MG
Encompass_Preferred
LUF5771 is a potent allosteric inhibitor of recombinant luteinizing hormone (recLH) and Org 43553. It can partially activate the LH receptor, though with low efficacy.
  • Acts as a potent allosteric inhibitor
  • Inhibits recombinant luteinizing hormone (recLH)
  • Targets and inhibits Org 43553
  • Capable of partial LH receptor activation
  • Exhibits low efficacy in LH receptor activation
Catalog No. 50-003-50353