Learn More
Medchemexpress LLC LUF5771 | 1141802-49-4 | 99.5% | 357.44 | 100 MG

Supplier: Medchemexpress LLC HY139303100MG
LUF5771 is a potent allosteric recombinant luteinizing hormone (recLH) and Org 43553 inhibitor. It partially activates the LH receptor with low efficacy. This compound is intended for research use to explore LH receptor function and modulation.
- Potent allosteric recLH and Org 43553 inhibitor
- Partially activates LH receptor with low efficacy
- Purity of 99.5%
- White to off-white solid appearance
- Soluble in DMSO and DMSO/corn oil mixture
- Specific storage conditions for powder and in solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.