Learn More
Medchemexpress LLC Lufenuron 10g | 511.15 g/mol | C17H8Cl2F8N2O3 | 10G
SDP
Supplier:  Medchemexpress LLC HY11558410G
Encompass_Preferred
Lufenuron is an analytical reference standard of the benzoylurea insecticide lufenuron, a lipophilic chitin synthesis inhibitor supplied for research and analytical use.
  • Analytical standard grade for reference and method development.
  • Used for qualitative and quantitative analysis by HPLC, GC, and MS.
  • Chitin synthesis inhibitor applicable to residue and parasitology studies.
  • Molecular weight 511.15 g/mol; formula C17H8Cl2F8N2O3; CAS 103055-07-8.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-28652