Learn More
Medchemexpress LLC Lufenuron Standard 100mg | 103055-07-8 | 511.15 g/mol | C17H8Cl2F8N2O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115584R100MG
Encompass_Preferred
Lufenuron is an analytical reference standard of a benzoylurea insecticide used as a chitin synthesis inhibitor. Supplied as a solid intended for analytical and methodological research applications, it includes batch-specific COA documentation and stated purity.
  • Purity 98.37% as stated on the certificate of analysis.
  • Suitable for HPLC, GC, and MS method development.
  • Available in small milligram pack sizes for analytical use.
  • Solid form, off-white to light yellow.
  • Store and handle according to the certificate of analysis instructions.
Catalog No. 50-002-28653