Learn More
OTT Hydromet Lufft - WS500 Smart Weather Sensor
Supplier:  OTT Hydromet 8373.U01
Encompass_Preferred
The WS500-UMB is a compact all-in-one weather sensor with measurement of temperature, relative humidity, air pressure, wind direction, wind speed. It is part of the WS product family of professional intelligent measuring sensors with a digital interface and an integrated design with ventilated radiation protection for various environmental applications. One external temperature or rain sensor is connectable.
Catalog No. NC1919625