Learn More
OTT Hydromet Lufft - Rain Gauge WTB100
Supplier:  OTT Hydromet 8353.1
Encompass_Preferred
Tipping bucket rain gauge with bounce-free reed contact. A 10 meter cable is included in scope of delivery.
Catalog No. NC1919633