Learn More
Apexbio Technology LLC Lumateperone tosylate 1187020-80-9 25mg
SDP
Supplier:  Apexbio Technology LLC A465525
Encompass_Preferred
Lumateperone tosylate (CAS 1187020-80-9) is a small molecule characterized as a selective antagonist of the 5-HT2A receptor (Ki 0 54 nM) It exhibits partial agonist activity at presynaptic dopamine D2 receptors and antagonist action at postsynaptic D2 receptors (Ki 32 nM) while also modulating dopamine D1 receptor function Preclinical studies indicate that lumateperone tosylate influences neurotransmitter release and synaptic signaling pathways It is utilized in research investigating the pathophysiology and treatment of neuropsychiatric disorders such as schizophrenia as well as in studies exploring its potential anticancer properties
Catalog No. 50-000-94946