Learn More
TARGETMOL CHEMICALS INC Lumefantrine 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3266100MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Lumefantrine (dl-Benflumelol) is an antimalarial agent used to treat acute uncomplicated malaria. It is administered in combination with artemether for improved efficacy. This combination therapy exerts its effects against the erythrocytic stages of Plasmodium spp. and may be used to treat infections caused by P. falciparum and unidentified Plasmodium species, including infections acquired in chloroquine-resistant areas. Purity 100%
Catalog No. 50-287-6315