Learn More
Analytical Sales & Services Lumidox II LumLamp
Supplier:  Analytical Sales & Services LUM2LWHTD80
Encompass
LumidoxII LumLamp, with WHITE LEDs, 80 Degree Extra Wide Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-06186