Learn More
Medchemexpress LLC Lumigolix | 1709823-61-9 | 98.6% | 607.57 | C28H27F2N9O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1348645MG
Encompass_Preferred
Lumigolix is a gonadotropin-releasing hormone (GnRH) receptor antagonist used in preclinical research, including endometriosis models. It is provided as a solid (white to yellow) and as a 10 mM solution in DMSO, with a molecular weight of 607.57 and high reported purity suitable for biological assays.
  • Selective GnRH receptor antagonist for endometriosis research.
  • High purity (98.58%), suitable for biochemical and cellular assays.
  • Physical form: solid, white to yellow; soluble in DMSO.
  • Available in small-scale solid quantities and as a ready-to-use DMSO solution.
  • Well characterized with reported molecular formula and weight for assay planning.
Catalog No. 50-002-23263