Learn More
Medchemexpress LLC HY-15922A 500mg Medchemexpress, Luminol (sodium salt) CAS:20666-12-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15922A500MG
Encompass_Preferred
Medchemexpress, HY-15922A 500mg Luminol (sodium salt) CAS:20666-12-0 Luminol sodium salt is a chemical that exhibits chemiluminescence with pKa values of 6.74 and 15.1. Luminol sodium salt exhibits chemiluminescence (CL) at 425 nm λmax. Luminol sodium salt is commonly used in forensics as a diagnostic tool for the detection of blood stains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1834