Learn More
Cayman Chemical LumogalIon 250mg
SDP
Supplier:  Cayman Chemical 35596250 MG
Encompass_Preferred
A fluorescent probe for aluminum; binds to aluminum, forming a complex that displays excitation/emission maxima of 500/590 nm, respectively; has been used to detect aluminum in natural waters and human urine and serum samples by HPLC, as well as in fixed soybean roots and THP-1 cells by confocal microscopy
Catalog No. 50-291-3447