Learn More
Medchemexpress LLC Lumogallion | 4386-25-8 | 99.8% | 344.73 | 100 MG

Supplier: Medchemexpress LLC HY137296100MG
Lumogallion is a highly sensitive fluorescent reagent used for the detection of aluminum, gallium, and other metals. It has an excitation wavelength of 490 nm and an emission spectrum ranging from 520 nm to 650 nm, with a peak near 580 nm. It can be used for detecting aluminum levels in brain tissue of Alzheimer's patients, staining live cells with intracellular aluminum adjuvant, and detecting aluminum content in plant root tip cells.
- Highly sensitive fluorescent reagent
- Detects aluminum, gallium, and other metals
- Excitation wavelength of 490 nm
- Emission spectrum from 520 nm to 650 nm, with a peak near 580 nm
- Suitable for detecting aluminum levels in brain tissue of Alzheimer's patients
- Can be used as a probe to stain live cells with intracellular aluminum adjuvant
- Detects aluminum content in root tip cell tissue of plants
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.