Learn More
Medchemexpress LLC Lumogallion | 4386-25-8 | MFCD00047770 | 99.79% | 344.73 | 50 MG

Supplier: Medchemexpress LLC HY13729650MG
Lumogallion is a highly sensitive fluorescent reagent used for the detection of aluminum, gallium, and other metals. It exhibits an excitation wavelength of 490 nm and an emission spectrum ranging from 520 nm to 650 nm, with a peak near 580 nm. This reagent is intended for research use only.
- Highly sensitive fluorescent reagent
- Used for detection of aluminum, gallium, and other metals
- Excitation wavelength of 490 nm
- Emission spectrum from 520 nm to 650 nm
- Peak emission near 580 nm
- Intended for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.