Learn More
NEW ENGLAND BIOGROUP LUNA 2nd General Reusable Slide, 2-pack, Counts 1 sample at a time, Contains LUNA reusable slide and one glass coverslip, For brightfield and fluorescent cell counts. One year warranty
SDP
Supplier:  NEW ENGLAND BIOGROUP L12012
Encompass
LUNA 2nd General Reusable Slide, 2-pack. Counts 1 sample at a time. Contains LUNA reusable slide and one glass coverslip. Can be used for brightfield and fluorescent cell counts. One year warranty
Catalog No. NC3903971