Learn More
Medchemexpress LLC Lunasin | 496823-34-8 | 95.6% | 5028.32 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP52855MG
Encompass_Preferred
Lunasin is a bioactive peptide derived from soybean, known for its antioxidant, anti-inflammatory, anticancer, and anti-aging properties. It operates through an epigenetic mechanism involving histone acetylation. Lunasin can be internalized by cells to inhibit Oncosphere formation in cancer cells. Its RGD motif facilitates internalization and contributes to its anti-transformation effects, particularly inhibiting metacercaria formation in melanoma cell lines.
  • Bioactive peptide with multiple therapeutic properties
  • Exhibits antioxidant effects
  • Demonstrates anti-inflammatory activity
  • Possesses anticancer and anti-aging properties
  • Isolated from soybean
  • Acts via epigenetic mechanism involving histone acetylation
  • Internalized by cells to inhibit oncosphere formation
  • RGD motif mediates cell internalization and anti-transformation effects
  • Inhibits metacercaria formation in melanoma cell lines
Catalog No. 50-003-99641