Learn More
Cellular Engineering Technologies Lung Cancer Associated Fibroblast Expansion Media (450mL)
Supplier:  Cellular Engineering Technologies MR1005
Encompass_Preferred
Lung cancer-associated fibroblast expansion media is designed to support the robust growth of LCAF cells. Media is compatible with available animal-origin serum, supplemented separately according to the customer preference.Media is shipped with gel packs.
Catalog No. NC2753066