Learn More
Medchemexpress LLC Lunresertib | 2719793-90-3 | 100.0% | C18H20N4O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY145817A10MM/1ML
Encompass_Preferred
Lunresertib (RP-6306) is a potent, selective, and orally active PKMYT1 inhibitor with an IC50 of 14 nM. It demonstrates high selectivity over other kinases in cellular binding assays and exhibits anticancer effects by inducing pan-γH2AX in HCC1569 breast cancer cell lines, making them vulnerable to DNA damage. Lunresertib also causes unscheduled activation of CDK1 in CCNE1-overexpressing cells, promoting early mitosis.
  • Potent and selective PKMYT1 inhibitor
  • Orally active with an IC50 of 14 nM
  • High selectivity over other kinases in cellular binding assays
  • Exhibits anticancer effects
  • Induces pan-γH2AX in HCC1569 breast cancer cell line
  • Causes unscheduled activation of CDK1 in CCNE1-overexpressing cells
Catalog No. 50-003-54038