Learn More
Medchemexpress LLC Lunularic acid | 23255-59-6 | MFCD24368585 | >=98.0% | 258.27 g·mol⁻1 | C15H14O4 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1245045MG
Encompass_Preferred
Lunularic acid is a dihydrostilbene compound used as an endogenous growth inhibitor in liverworts and supplied for research use. It shows anti-hyaluronidase activity (IC50: 0.13 nM) and is provided as an off-white solid for biochemical and plant physiology studies.
  • Acts as an endogenous growth inhibitor in liverworts.
  • Exhibits anti-hyaluronidase activity (IC50: 0.13 nM).
  • Molecular formula C15H14O4; molecular weight 258.27 g·mol⁻1.
  • Off-white solid form suitable for laboratory assays.
  • Commonly supplied in milligram-scale quantities for research applications.
Catalog No. 50-002-36132