Learn More
Medchemexpress LLC Lunularic acid | 23255-59-6 | MFCD24368585 | 99.2% | 258.27 g/mol | C15H14O4 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1245041MG
Encompass_Preferred
Lunularic acid is a dihydrostilbene small-molecule research compound reported as an endogenous growth inhibitor with anti-hyaluronidase activity. It is used in biochemical and pharmacological studies of plant growth regulation and enzyme inhibition, and is typically supplied in small milligram quantities for laboratory research.
Catalog No. 50-002-39333