Learn More
Enzo Life Sciences Lupeol (10mg). CAS: 545-47-1
SDP
Supplier:  Enzo Life Sciences ALX350355M010
Encompass_Preferred
Anti-inflammatory, antioxidant and antimutagenic compound. Shows preventive effects on DMBA induced DNA alkylation damage in mouse skin. Modulator of NF-κB and PI(3)K/Akt (protein kinase B; PKB) pathways. Induces Fas-mediated apoptosis via inhibition of Ras signalling. Inhibits tumor growth in mouse in vivo. Alternative name: Fagarasterol, 20(29)-Lupen-3beta-ol, 3beta-Hydroxy-20(29)-lupene. Purity: ≥98% (HPLC). Solubility: Soluble in ether or warm 100% ethanol; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8858