Learn More
Medchemexpress LLC HY-B0032A 100mg Medchemexpress, Lurasidone CAS:367514-87-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYB0032A100MG
Encompass_Preferred
Medchemexpress,HY-B0032A 100mg Lurasidone CAS:367514-87-2 Formula:C28H36N4O2S Lurasidone is an antagonist of both dopamine D2 and 5-HT7 with IC50s of 1.68 and 0.495 nM, respectively. Lurasidone is also a partial agonist of 5-HT1A receptor with an IC50 of 6.75 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9151