Learn More
Medchemexpress LLC Lurasidone-d8-1 hydrochloride | 367514-88-3 | 537.19 g/mol | C28H29D8ClN4O2S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6540101MG
Encompass_Preferred
Lurasidone-d8-1 hydrochloride is a deuterium-labeled analogue of lurasidone hydrochloride supplied as a research-grade reference standard for analytical and metabolic studies. It is intended as an isotopically labeled internal standard for LC-MS/MS, ADME, and bioanalytical method development, and is provided in small-scale quantities for laboratory use. This product is for research use only and not for human or clinical use.
  • Deuterium-labeled analogue of lurasidone hydrochloride.
  • Suitable as an isotopically labeled internal standard for LC-MS/MS and quantitative analysis.
  • Intended for research use only, not for human or clinical use.
  • Available in small-scale quantities appropriate for analytical assays.
  • Molecular formula: C28H29D8ClN4O2S; molecular weight: 537.19 g/mol.
Catalog No. 50-002-84241