Learn More
Selleck Chemical LLC Lurasidone HCl S3044-25mg
SDP
Supplier:  Selleck Chemical LLC S304425MG
Encompass_Preferred
Lurasidone (SM-13496) is an atypical antipsychotic inhibits Dopamine D2 5-HT2A 5-HT7 5-HT1A and noradrenaline 2C with IC50 of 1 68 nM 2 03 nM 0 495 nM 6 75 nM and 10 8 nM respectively
Catalog No. 50-311-0008