Learn More
Apexbio Technology LLC Lurasidone HCl 367514-88-3 50mg
SDP
Supplier:  Apexbio Technology LLC B147750
Encompass_Preferred
Lurasidone HCl (CAS 367514-88-3) is an atypical antipsychotic small molecule exhibiting potent inhibitory activity against multiple neurotransmitter receptors It acts as an antagonist of dopamine D2 serotonin 5-HT2A 5-HT7 5-HT1A and noradrenergic 2C receptors with IC50 values of 1 68 nM 2 03 nM 0 495 nM 6 75 nM and 10 8 nM respectively Through modulation of these receptor pathways lurasidone hydrochloride influences neurotransmission involved in psychiatric disorders making it particularly relevant for neuropharmacological research on schizophrenia and related conditions
Catalog No. 50-000-62022