Learn More
Medchemexpress LLC Lurasidone (Hydrochloride) | 367514-88-3 | 99.65% | 529.14 | 500 MG
SDP
Supplier:  Medchemexpress LLC 500MGHYB0032
Encompass_Preferred
Lurasidone (Hydrochloride) acts as an antagonist for dopamine D2 and 5-HT7 receptors, with IC50s of 1.68 nM and 0.495 nM, respectively. It also functions as a partial agonist for the 5-HT1A receptor, exhibiting an IC50 of 6.75 nM.
  • Shows higher affinity for dopamine D2 and 5-HT2A receptors compared to other antipsychotics tested.
  • Dose-dependently increases the DOPAC/dopamine ratio in both the frontal cortex and striatum, with a more pronounced effect on the frontal cortex at higher doses.
  • Dose-dependently inhibits conditioned avoidance response (CAR) in rats.
  • Dose-dependently inhibits TRY-induced forepaw clonic seizure and p-CAMP-induced hyperthermia.
Catalog No. 50-003-65725