Learn More
Medchemexpress LLC Lurasidone metabolite 14326 hydrochloride | 99.4% | 545.14 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYG0002A1MG
Encompass_Preferred
Lurasidone metabolite 14326 hydrochloride is an active metabolite of the atypical antipsychotic Lurasidone. Intended for research use only, this product is not sold to patients. While the salt and free forms of the compound exhibit comparable biological activity at equivalent molar concentrations, the hydrochloride salt form typically offers enhanced water solubility and stability.
  • Active metabolite of atypical antipsychotic lurasidone
  • Intended for research use only
  • Enhanced water solubility compared to free base
  • Enhanced stability compared to free base
  • High purity of 99.42%
Catalog No. 50-004-29899