Learn More
Alkali Scientific Luria Broth, High Salt Formula, Powder [Miller's LB Broth], 1 Kg
SDP
Supplier:  Alkali Scientific LUA001CP
Encompass
Luria Broth (High Salt Formula) Powder is a microbial growth medium used for cultivating bacteria that require higher sodium concentrations. This medium comes in a powder form and is easily rehydrated with water. The 1kg size is ideal for small to medium-scale laboratories or research projects. The high salt concentration enhances the growth of specific bacterial strains that thrive in saline environments, making it suitable for certain types of microbiological research. Luria Broth is commonly used for culturing Escherichia coli and other bacteria in molecular biology, microbiology, and biotechnology applications, including plasmid cloning and protein expression.
Catalog No. 50-006-60577