Learn More
Alkali Scientific Luria Broth [Miller's LB] 1 Liter Buffered Capsules, 40 Capsules of 25 Grams, 1 Kg
SDP
Supplier:  Alkali Scientific ABR001CP
Encompass_Preferred
Luria Broth [Miller's LB] 1 Liter Buffered Capsules, 40 Capsules of 25 Grams, 1 Kg is a versatile culture medium used for the growth of microorganisms in various microbiological applications. The buffered formula ensures that the pH remains stable during incubation, which is essential for the reliable growth of a wide range of bacterial and yeast species. The 1-liter volume format is perfect for laboratories that need a larger amount of media for microbial culture or other laboratory experiments. These capsules are designed for ease of use, dissolving quickly in water to provide a consistent and reliable media solution for bacterial cultivation, DNA research, and protein expression.
Catalog No. 50-006-61530