Learn More
Medchemexpress LLC Luspatercept 50mg | 50mg
SDP
Supplier:  Medchemexpress LLC HYP9972050MG
Encompass_Preferred
Luspatercept (ACE-536) is a recombinant modified ActRIIB fusion protein that binds with transforming growth factor superfamily ligands Luspatercept increases the erythrocyte numbers and promotes maturation of erythroid precursors Luspatercept binds with GDF11 and inhibits Smad2/3 signaling Luspatercept can be used for the research of anemia[1]
Catalog No. 50-003-22286
Add to Cart
Request a Quote