Learn More
Medchemexpress LLC Luteinizing Hormone beta Antibody (YA1095) | 50 UL

Supplier: Medchemexpress LLC HYP8135050UL
Luteinizing Hormone beta Antibody (YA1095) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Luteinizing Hormone beta. It is for research use only.
- Rabbit-derived IgG monoclonal antibody
- Targets Luteinizing Hormone beta
- Predicted band size: 16 kDa
- Observed band size: 19 kDa
- Species reactivity: Human
- Purity: Affinity purified
- Non-conjugated
- Formulation: Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.