Learn More
Alpco Ltd Luteinizing Hormone ELISA (LH ELISA)
SDP
Supplier:  Alpco Ltd 11LUTHUE01
Encompass
The Luteinizing Hormone ELISA (LH ELISA) is for the direct quantitative determination of Luteinizing Hormone by enzyme immunoassay in human serum. FDA Registered. For In Vitro Diagnostic Use in the United States.
Catalog No. 50-921-698