Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luteolin 7-diglucuronide | 96400-45-2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN72695MG

Encompass_Preferred

Luteolin 7-diglucuronide is the major flavonoid isolated from *Aloysia triphylla* and *Verbena officinalis*. This product is for research use only.

  • Purity: 99.34%
  • Molecular weight: 638.48
  • Formula: C27H26O18
  • Appearance: Solid
  • Color: Yellow to brown
  • Structure classified as flavonoids, flavones, phenols, and polyphenols
  • Initial source from plants (other families)
  • Inhibits activity of protein tyrosine phosphatase 1B (PTP1B)
  • Upregulates phosphorylation of AMPK
  • Inhibits activation of hepatic stellate cells
  • Exhibits protective effects against isoproterenol-induced myocardial injury and fibrosis
  • Reduces myocardial necrosis and inflammation
  • Inhibits expression of NADPH oxidase subunits and ECM genes

Catalog No. 50-004-19483


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.