Learn More
Selleck Chemical LLC Luteolin 50mg 491-70-3
SDP
Supplier:  Selleck Chemical LLC S232050MG
Encompass_Preferred
Luteolin is a flavonoid found in Terminalia chebula, which is a non-selective phisphodiesterase PDE inhibitor for PDE1-5 with Ki of 15.0 μM, 6.4 μM, 13.9 μM, 11.1 μM and 9.5 μM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-667-5