Learn More
Medchemexpress LLC Luvixasertib hydrochloride | 1610677-37-6 | 99.6% | 535.04 g/mol | C28H31ClN6O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101340A100MG
Encompass_Preferred
Luvixasertib hydrochloride is a selective, orally bioavailable inhibitor of the TTK/Mps1 kinase with potent in vitro activity (IC50 = 1.7 nM) and demonstrated antitumor efficacy in preclinical models. Supplied as a light-yellow solid hydrochloride salt for research use, the compound is characterized by high purity and defined storage recommendations.
  • Selective TTK/Mps1 kinase inhibition
  • Potent in vitro activity with IC50 of 1.7 nM
  • Demonstrated antitumor activity in xenograft and patient-derived models
  • High reported purity (99.63%)
  • Supplied as a light-yellow solid suitable for formulation or dissolution
  • Available in multiple pack sizes, including 100 mg
  • Storage: solid at 4°C; in solution -80°C for long-term stability
Catalog No. 50-002-13203