Learn More
Imanis Life Sciences LV-SFFV-LUC2-P2A-EMGFP (1ML)
Supplier:  Imanis Life Sciences LV050L
Encompass
LV-SFFV-Luc2-P2A-EmGFP (1mL)PROMOT -Firefly luciferase (Luc2)Emerald green fluorescent protein (EmGFP)PROMOTER-Spleen focus-forming virus (SFFV)This is a ready-to-use lentivirus preparation. The virus encodes the firefly luciferase (Luc2) cDNA linked to the emerald green fluorescent protein (EmGFP) cDNA via a P2A cleavage peptide; the spleen focus-forming virus (SFFV) promoter transcriptionally controls expression of the Luc2-P2A-EmGFP cDNA.
Catalog No. NC1840215