Learn More
Apexbio Technology LLC LW6 934593-90-5 5mg
SDP
Supplier:  Apexbio Technology LLC A47655
Encompass_Preferred
LW6 (CAS 934593-90-5) is a small-molecule inhibitor that selectively suppresses hypoxia-inducible factor 1-alpha (HIF-1 ) protein expression without affecting HIF-1 levels exhibiting an IC50 of approximately 4 4 M It also demonstrates inhibitory activity against malate dehydrogenase 2 (MDH2) Given the role of HIF-1 in cellular adaptation to hypoxia and angiogenesis LW6 serves as a valuable tool for investigating HIF-related signaling pathways hypoxic responses and for developing therapeutic strategies targeting diseases associated with aberrant hypoxia signaling
Catalog No. 50-000-93529